Läs mer
Bacitracin A (Technical Grade), TRC
Produktkod. 30331331
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30331331 5 mg
1 options
Produktkod. 30331331 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1065335MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bacitracin A
CAS 22601-59-8
Molekylformel C66H103N17O16S
Kvantitet 5 mg
Synonym Bacitracin A1, 1-[N-[[2-(1-amino-2-methylbutyl)-4,5-dihydro-4-thiazolyl]carbonyl]-L-leucine]-Bacitracin F, 1,4,7,10,13,16,19-Heptaazacyclopentacosane, cyclic peptide deriv., Bacitracin A2a, NSC 45737
InChI formel InChI=1S/C66H103N17O16S/c1-9-35(6)52(69)66-81-48(32-100-66)63(97)76-43(26-34(4)5)59(93)74-42(22-23-50(85)86)58(92)83-53(36(7)10-2)64(98)75-40-20-15-16-25-71-55(89)46(29-49(68)84)78-62(96)47(30-51(87)88)79-61(95)45(28-39-31-70-33-72-39)77-60(94)44(27-38-18-13-12-14-19-38)80-65(99)54(37(8)11-3)82-57(91)41(21-17-24-67)73-56(40)90/h12-14,18-19,31,33-37,40-48,52-54H,9-11,15-17,20-30,32,67,69H2,1-8H3,(H2,68,84)(H,70,72)(H,71,89)(H,73,90)(H,74,93)(H,75,98)(H,76,97)(H,77,94)(H,78,96)(H,79,95)(H,80,99)(H,82,91)(H,83,92)(H,85,86)(H,87,88)/t35-,36-,37-,40-,41+,42+,43-,44+,45-,46-,47+,48-,52-,53-,54-/m0/s1
LEDER CC[C@@H]([C@H](N)C1=N[C@H](C(N[C@H](C(N[C@@H](C(N[C@H](C(N[C@H]2CCCCNC([C@@H](NC([C@H](NC([C@@H](NC([C@H](NC([C@@H](NC([C@H](NC2=O)CCCN)=O)[C@H](CC)C)=O)CC3=CC=CC=C3)=O)CC4=CN=CN4)=O)CC(O)=O)=O)CC(N)=O)=O)=O)[C@H](CC)C)=O)CCC(O)=O)=O)CC(C)C)=O)CS1)C
IUPAC-namn (4R)-4-[[(2S)-2-[[(4R)-2-[(1S,2S)-1-amino-2-methylbutyl]-4,5-dihydro-1,3-thiazole-4-carbonyl]amino]-4-methylpentanoyl]amino]-5-[[(2S,3S)-1-[[(3S,6R,9S,12R,15S,18R,21S)-3-(2-amino-2-oxoethyl)-18-(3-aminopropyl)-12-benzyl-15-[(2S)-butan-2-yl]-6-(carboxymethyl)-9-(1H-imidazol-4-ylmethyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclopentacos-21-yl]amino]-3-methyl-1-oxopentan-2-yl]amino]-5-oxopentanoic acid
Molekylvikt (g/mol) 1422.69
Formel vikt 1421.7489