Läs mer
Bacitracin F (~75%), TRC
Produktkod. 17952535
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17952535 10 mg
17974735 250 mg
17984245 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17952535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB10653010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22601-63-4
Molekylformel C66 H98 N16 O17 S
Molekylvikt (g/mol) 1419.65
Synonym L-Asparagine, N-[[2-[(2S)-2-methyl-1-oxobutyl]-4-thiazolyl]carbonyl]-L-leucyl-D-α-glutamyl-L-isoleucyl-L-lysyl-D-ornithyl-L-isoleucyl-D-phenylalanyl-L-histidyl-D-α-aspartyl-, (10→4)-lactam, Bacitracin F (8CI,9CI), 1,4,7,10,13,16,19-Heptaazacyclopentacosane, cyclic peptide deriv., 4,10-Anhydro[N-[[2-[(2S)-2-methyl-1-oxobutyl]-1,3-thiazol-4-yl]carbonyl]-L-leucyl-D-α-glutamyl-L-isoleucyl-L-lysyl-D-ornithyl-L-isoleucyl-D-phenylalanyl-L-histidyl-D-α-aspartyl-L-asparagine]
IUPAC-namn (4R)-5-[[(2S,3S)-1-[[(3S,6R,9S,12R,15S,18R,21S)-3-(2-amino-2-oxoethyl)-18-(3-aminopropyl)-12-benzyl-15-[(2S)-butan-2-yl]-6-(carboxymethyl)-9-(1H-imidazol-4-ylmethyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclopentacos-21-yl]amino]-3-methyl-1-oxopentan-2-yl]amino]-4-[[(2S)-4-methyl-2-[[2-[(2S)-2-methylbutanoyl]-1,3-thiazole-4-carbonyl]amino]pentanoyl]amino]-5-oxopentanoic acid
LEDER CC[C@H](C)[C@H](NC(=O)[C@@H](CCC(=O)O)NC(=O)[C@H](CC(C)C)NC(=O)c1csc(n1)C(=O)[C@@H](C)CC)C(=O)N[C@H]2CCCCNC(=O)[C@H](CC(=O)N)NC(=O)[C@@H](CC(=O)O)NC(=O)[C@H](Cc3c[nH]cn3)NC(=O)[C@@H](Cc4ccccc4)NC(=O)[C@@H](NC(=O)[C@@H](CCCN)NC2=O)[C@@H](C)CC
Specifikationer
Kemiskt namn eller material Bacitracin F
Analyt- eller komponentnamn Bacitracin F
Frakt skick Room Temperature
InChI formel InChI=1S/C66H98N16O17S/c1-9-35(6)52(82-58(92)42(22-23-50(84)85)73-59(93)43(26-34(4)5)75-63(97)48-32-100-66(80-48)54(88)37(8)11-3)64(98)74-40-20-15-16-25-70-55(89)46(29-49(68)83)77-62(96)47(30-51(86)87)78-61(95)45(28-39-31-69-33-71-39)76-60(94)44(27-38-18-13-12-14-19-38)79-65(99)53(36(7)10-2)81-57(91)41(21-17-24-67)72-56(40)90/h12-14,18-19,31-37,40-47,52-53H,9-11,15-17,20-30,67H2,1-8H3,(H2,68,83)(H,69,71)(H,70,89)(H,72,90)(H,73,93)(H,74,98)(H,75,97)(H,76,94)(H,77,96)(H,78,95)(H,79,99)(H,81,91)(H,82,92)(H,84,85)(H,86,87)/t35-,36-,37-,40-,41+,42+,43-,44+,45-,46-,47+,52-,53-/m0/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 10 mg
Formel vikt 1418.7017