Learn More
CPC™ Back-to-Back Insert Adapter
Product Code. 17874454
Change view
Click to view available options
Pressure:
Up to 60 psi, 4.1 bar
Unit Size:
Pack of 25
Product Code. Pressure Unit Size
17874454 Up to 60 psi, 4.1 bar Pack of 25
1 options
Product Code. 17874454 Supplier CPC™ Supplier No. MPC22X2239M
 Restricted Item
Beräknad leverans: 18-01-2027
MPC to MPX Back-to-Back Insert Adapter
  • Medical Grade
  • Non-valved
  • Seal Options: Silicone, platinum-cured, USP Class VI, ADCF
Specifications
Certifications/Compliance USP Class VI, ADCF
Color Amber
Material Polysulfone
Pressure Up to 60 psi, 4.1 bar
Sterilization Method Gamma, Autoclave
Temperature (English) Max. 300°F
Temperature (English) Min -40°F
Temperature (Metric) Max 149°C