missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Bad Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56079
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Bad Polyclonal specifically detects Bad in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifikationer
| Bad | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Bad | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| BAD | |
| A synthetic peptide corrsponding to amino acids 2-23 (GTPKQPSLAPAHALGLRKSDPG) of mouse/rat Bad was used as immunogen; GenBank nos. NP_031548.1/NP_073189.1. | |
| 0.05 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 572 | |
| Mouse, Rat | |
| Antisera |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering