Läs mer
Invitrogen™ BAGE2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567594
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
BAGE2 Polyclonal Antibody for Western Blot
BAGE2 (BAGE Family Member 2) is a Protein Coding gene. Candidate gene encoding tumor antigens.
Specifikationer
| BAGE2 | |
| Polyclonal | |
| Unconjugated | |
| BAGE2 | |
| B melanoma antigen 2; B melanoma antigen family member 2; B melanoma antigen family, member 2; BAGE2; cancer/testis antigen 2.2; cancer/testis antigen family 2, member 2; CT2.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85319 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q86Y30 | |
| BAGE2 | |
| Synthesized peptide derived from human BAGE2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |