Learn More
Interscience™ BagOpen 400 for DiluFlow
Product Code. 17545245
Change view
Click to view available options
For Use With (Equipment):
DiluFlow, DiluFlow Pro, DiluFlow Elite
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17545245 DiluFlow, DiluFlow Pro, DiluFlow Elite Each
1 options
Product Code. 17545245 Supplier Interscience™ Supplier No. 505016
  • ONLINE EXCLUSIVE PRICE
    8395.00 SEK
    7496.88 SEK / Paket med 1
    Save 898.12 SEK
    10% Av
 Restricted Item
Beräknad leverans: 07-01-2027
BagOpen 400 for dilutor.
  • Made of durable stainless steel
Specifications
Product Type Bag Stand
Autoclavable Not Autoclavable
Volume (Metric) 400 mL
For Use With (Application) Standing Up Bags
For Use With (Equipment) DiluFlow, DiluFlow Pro, DiluFlow Elite