Läs mer
Baicalein Monohydrate, TRC
Produktkod. 30339219
Change view
Kvantitet:
500 mg
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30339219 10 g
30331169 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30339219 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB11525010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Baicalein, Monohydrate
CAS 352000-07-8
Molekylformel C15H12O6
Kvantitet 10 g
Synonym 5,6,7-Trihydroxyflavone Monohydrate, 5,6,7-Trihydroxy-2-phenyl-4H-1-benzopyran-4-one Hydrate
InChI formel InChI=1S/C15H10O5.H2O/c16-9-6-11(8-4-2-1-3-5-8)20-12-7-10(17)14(18)15(19)13(9)12;/h1-7,17-19H;1H2
LEDER O.Oc1cc2OC(=CC(=O)c2c(O)c1O)c3ccccc3
IUPAC-namn 5,6,7-trihydroxy-2-phenylchromen-4-one;hydrate
Molekylvikt (g/mol) 288.26
Formel vikt 288.0634