Läs mer
Tocris Bioscience™ Balicatib
Potent and selective cathepsin K inhibitor
Brand: Tocris Bioscience™ 5585/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Balicatib is a potent and selective cathepsin K inhibitor (IC50 values are 1.4, 56 and 480 nM for human, rat and mouse cathepsin K, respectively). Exhibits >300-fold selectivity for cathepsin K over cathepsins L, B and S. Long term supratherapeutic dosing increases tissue levels of cathepsins L and B in rats.Specifikationer
| N-[1-(Cyanomethylcarbamoyl)cyclohexyl]-4-(4-propylpiperazin-1-yl)benzamide | |
| 354813-19-7 | |
| 10 mg |