Läs mer
Balipramine Hydrochloride (Impurity), TRC
Produktkod. 30346613
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30346613 1 g
1 options
Produktkod. 30346613 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1157001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Balipramine Hydrochloride (Impurity)
CAS 58262-51-4
Molekylformel C19H22N2 . HCl
Kvantitet 1 g
Synonym N,N-Dimethyl-5H-dibenz[b,f]azepine-5-propanamine Hydrochloride, G 31406 Hydrochloride, 10,11-Dehydroimipramine Hydrochloride, Depramine Hydrochloride
InChI formel InChI=1S/C19H22N2.ClH/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21;/h3-6,8-13H,7,14-15H2,1-2H3;1H
LEDER Cl.CN(C)CCCN1c2ccccc2C=Cc3ccccc13
IUPAC-namn 3-benzo[b][1]benzazepin-11-yl-N,N-dimethylpropan-1-amine;hydrochloride
Molekylvikt (g/mol) 314.9
Formel vikt 314.155