Läs mer
Balsalazide, TRC
Produktkod. 30330220
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30330220 5 mg
1 options
Produktkod. 30330220 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1163005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Balsalazide
CAS 80573-04-2
Molekylformel C17 H15 N3 O6
Kvantitet 5 mg
Synonym Benzoic acid, 5-[(1E)-2-[4-[[(2-carboxyethyl)amino]carbonyl]phenyl]diazenyl]-2-hydroxy-, Benzoic acid, 5-[(1E)-[4-[[(2-carboxyethyl)amino]carbonyl]phenyl]azo]-2-hydroxy- (9CI), Benzoic acid, 5-[[4-[[(2-carboxyethyl)amino]carbonyl]phenyl]azo]-2-hydroxy-, (E)-, 5-[(1E)-2-[4-[[(2-Carboxyethyl)amino]carbonyl]phenyl]diazenyl]-2-hydroxybenzoic acid, Balsalazida, Balsalazide, Balsalazido, Balsalazidum, Colaza, Colazide, Giazo
InChI formel InChI=1S/C17H15N3O6/c21-14-6-5-12(9-13(14)17(25)26)20-19-11-3-1-10(2-4-11)16(24)18-8-7-15(22)23/h1-6,9,21H,7-8H2,(H,18,24)(H,22,23)(H,25,26)/b20-19+
LEDER OC(=O)CCNC(=O)c1ccc(cc1)N=Nc2ccc(O)c(c2)C(=O)O
IUPAC-namn 5-[(E)-[4-(2-carboxyethylcarbamoyl)phenyl]diazenyl]-2-hydroxybenzoic acid
Molekylvikt (g/mol) 357.32
Formel vikt 357.0961