Läs mer
Tocris Bioscience™ BAM 15
GREENER_CHOICE
Produktkod. 18756019
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18756019 10 mg
18725788 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18756019 Leverantör Tocris Bioscience™ Leverantörsnummer 5737/10
Mitochondrial protonophore uncoupler
BAM 15 is a mitochondrial protonophore uncoupler; uncouples oxidative phosphorylation in L6 myoblast mitochondria in vitro (EC50 = 270 nM). Does not depolarize the plasma membrane. Protects mice from acute renal ischemic-reperfusion injury.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N5,N6-bis(2-Fluorophenyl)-[1,2,5]oxadiazolo[3,4-b]pyrazine-5,6-diamine
CAS 210302-17-3
Kvantitet 10 mg
Mål Oxidative Phosphorylation Inhibitors
Molekylformel C16H10F2N6O
Formel vikt 340.29
Renhet >98%