missing translation for 'onlineSavingsMsg'
Learn More

BAR501, MedChemExpress

Product Code. 16486037
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
16486037 5 mg -
16463318 10 mg -
16454566 100 mg 100mg
16440749 1 mg -
16439856 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16486037 Supplier Medchem Express Supplier No. HY101274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAR501 is a potent and selective agonist of GPBAR1 with an EC50 of 1 μM.

BAR501 is a potent and selective agonist of GPBAR1 with an EC50 of 1 μM.

Specifications

Kemiskt namn eller material BAR501
CAS 1632118-69-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C26H46O3
Kvantitet 5 mg
Löslighetsinformation Ethanol : 120 mg/mL (295.10 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (122.96 mM)
LEDER C[C@H](CCCO)[C@@]1([H])CC[C@@]2([H])[C@]3([H])[C@@H](O)[C@@H](CC)[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])CC[C@@]21C
Molekylvikt (g/mol) 406.64
Formel vikt 406.64
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.