missing translation for 'onlineSavingsMsg'
Learn More

Barium Carbonate, 99.0 to 101.0%, ACS Reagent, Honeywell™ Fluka™

Catalog Number p-7120207
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
513-77-9
CBaO3
197.34
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
10563
[Ba++].[O-]C([O-])=O
This item is not returnable. View return policy
513-77-9
CBaO3
197.34
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
10563
[Ba++].[O-]C([O-])=O

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 1400°C
Color Colorless or White
pH 7 to 8 at 20°C
Quantity 500 g
Linear Formula BaCO3
UN Number NONH for all modes of transport
Beilstein 7045119
IUPAC Name barium(2+);carbonate
Formula Weight 197.34g/mol
Percent Purity 99.0 to 101.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Barium Carbonate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'