Läs mer
Barium Chloride Solution R1 - British Pharmacopoeia, Fisher Chemical™
Produktkod. 30695067 Handla allt Fisher Chemical Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30695067 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 30695067 Leverantör Fisher Chemical Leverantörsnummer BPR 047
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Pharmacopoeia reagents for testing and monographs, produced under a GMP-compliant ISO 9001 quality management system.
    This Pharmacopoeia reagent is manufactured :
  • Using pharmacopoeia reagent quality components
  • Water meeting the relevant pharmacopoeia monograph requirement for Purified Water
  • Supplied with a Certificate of Analysis for Performance Test
  • Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO 17034
Hazard Statement: May be flammable or oxidizing; may cause severe skin burns and serious eye damage or irritation; may cause respiratory irritation, drowsiness or dizziness; may be toxic if swallowed, in contact with skin, or if inhaled; may cause cancer, genetic defects, reproductive toxicity or developmental harm; may be harmful to aquatic life with long-lasting effects; may be corrosive to metals.
Specifikationer
Kvantitet 1000 mL