Learn More
Sherwood Scientific™ Barium Filter
Brand:  Sherwood Scientific™ 40016005
Additional Details : Weight : 1.00000kg
Product Code. 12911821
  • 3230.00 SEK / Each
Estimated Shipment: 30-08-2024
to see stock.
Specifications
M410 Flame Photometer
Barium Filter