Läs mer
Barium Hydroxide Solution - British Pharmacopoeia, Fisher Chemical™
Produktkod. 30694991 Handla allt Fisher Chemical Produkter
Change view
Kvantitet:
1000 mL
Produktkod. Kvantitet
30694991 1000 mL
1 options
Produktkod. 30694991 Leverantör Fisher Chemical Leverantörsnummer BPR 049
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Pharmacopoeia reagents for testing and monographs, produced under a GMP-compliant ISO 9001 quality management system.
    This Pharmacopoeia reagent is manufactured :
  • Using pharmacopoeia reagent quality components
  • Water meeting the relevant pharmacopoeia monograph requirement for Purified Water
  • Supplied with a Certificate of Analysis for Performance Test
  • Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO 17034
Hazard Statement: May be flammable or oxidizing; may cause severe skin burns and serious eye damage or irritation; may cause respiratory irritation, drowsiness or dizziness; may be toxic if swallowed, in contact with skin, or if inhaled; may cause cancer, genetic defects, reproductive toxicity or developmental harm; may be harmful to aquatic life with long-lasting effects; may be corrosive to metals.
Specifikationer
Kvantitet 1000 mL