Läs mer
Barlerin, MedChemExpress
Produktkod. 16473896
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16473896 25 mg
16451498 5 mg
16439088 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16473896 Leverantör Medchem Express Leverantörsnummer HYN0758
MedChemExpress Barlerin (8-O-Acetyl shanzhiside methyl ester) is an iridoid glucoside isolated from the leaves of Lamiophlomis rotata Kudo, a Chinese folk medicinal plant in Xi-zang. Barlerin (8-O-Acetyl shanzhiside methyl ester) could inhibt NF-κB.
Barlerin (8-O-Acetyl shanzhiside methyl ester) is an iridoid glucoside isolated from the leaves of Lamiophlomis rotata Kudo, a Chinese folk medicinal plant in Xi-zang. Barlerin (8-O-Acetyl shanzhiside methyl ester) could inhibt NF-κB.
Specifikationer
Kemiskt namn eller material Barlerin
CAS 57420-46-9
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C19H28O12
Kvantitet 25 mg
Synonym 8-O-Acetyl shanzhiside methyl ester
Löslighetsinformation DMSO : 100 mg/mL (223.01 mM; Need ultrasonic)
LEDER O[C@H]([C@@H](O)[C@@H]1O)[C@](O[C@@H]1CO)([H])O[C@H]2[C@@]([C@@]3(C)OC(C)=O)([H])[C@@]([C@H](O)C3)([H])C(C(OC)=O)=CO2
Molekylvikt (g/mol) 448.42
Formel vikt 448.42
Procent renhet 99.0%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Neuroscience-Neuromodulation