Läs mer
Daniels Healthcare™ Mini Bin Sharps Container Base
:
For Use With: Mini Sharps Bin / Bin Top
Beskrivning
- Designed to meet demands for strength, simplicity and value.
- Containers offer outstanding protection against penetrating sharps and leaking fluids due to the rigid polypropylene construction and wall thickness.
- Containers are manufactured to BS 7320:1990, ISO 9002 standards and UN Packing Group II approved.
- Containers have all British Standard markings, user instructions and a visible fill line
Specifikationer
Specifikationer
| För användning med (applikation) | Safe disposal of sharps |
| För användning med (utrustning) | Mini Sharps Bin / Bin Top |