Läs mer
Tork Basic Paper W6, Wiping Paper Towel Roll
Handla allt Tork Produkter
Färg:
Blått
Vitt
Beskrivning
- Efficiently clean, even in high humidity and wet environments, with this specially designed wiping paper.
- Ensure you have high-quality wiping paper, thanks to these food contact certified rolls.
- Making your environment HACCP compliant is made easier with these HACCP International certified rolls.
- Improve food production safety, thanks to the high visibility and traceability of this paper roll.
- 6 x 250 m roll
Specifikationer
Specifikationer
| Produkttyp | Wiping Paper |
| Längd (engelska) | 273.4 yd. |
| Längd (metrisk) | 250 m |
| Bredd (engelska) | 7.68 in. |
| Bredd (metrisk) | 19.5 cm |
| Autoklaveringsbar | Not Autoclavable |
| Färg | Vitt |
| Förpackningsmängd | 1 Roll |
| Lager | 1-ply |