Läs mer
Basic Red 51 (Technical Grade), TRC
Produktkod. 30336677
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30336677 50 mg
1 options
Produktkod. 30336677 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB11874050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Basic Red 51 (Technical Grade)
CAS 12270-25-6
Molekylformel C13H18ClN5
Kvantitet 50 mg
Synonym Basacryl Red X-BL, Basic Red 51, Maxilon Red M-RL, Maxilon Red SL, Maxilon Red SL 200, Vibracolor Ruby Red
InChI formel InChI=1S/C13H18N5.ClH/c1-16(2)12-7-5-11(6-8-12)14-15-13-17(3)9-10-18(13)4;/h5-10H,1-4H3;1H/q+1;/p-1
LEDER CN1C=C[N+](=C1N=NC2=CC=C(C=C2)N(C)C)C.[Cl-]
IUPAC-namn 2-((4-(dimethylamino)phenyl)diazenyl)-1,3-dimethyl-1H-imidazol-3-ium chloride
Molekylvikt (g/mol) 279.77
Formel vikt 279.13