missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Battery Charger for Technotrate Digital Burette
Kartell™ Battery Charger for Technotrate Digital Burette is a crucial accessory to be used with a digital burette.
Brand: Kartell™ 8302100
Additional Details : Weight : 5.00000kg
Description
- For Technotrate Digital Burette
- Electrical requirements of 220V and 50Hz
- Battery charger
Specifications
Battery Charger | |
220V, 50Hz | |
Technotrate Digital Burette |