Läs mer
Tocris Bioscience™ Bax channel blocker
Kvantitet:
10 mg
50 mg
Beskrivning
Bax channel blocker is a allosteric inhibitor of Bax channel activation. Binds inactive Bax at allosteric site and inhibits tBID-mediated Bax activation (IC50 = 3.3 μM). Selectively inhibits Bax-dependent apoptosis. Potent inhibitor of Bax-mediated mitochondrial cytochrome c release (IC50 = 0.52 μM).
Specifikationer
Specifikationer
| Kvantitet | 50 mg |
| Produkttyp | Bax channel blocker |
| Renhet | >98% |
| Molekylvikt (g/mol) | 540.12 |
| Inhibitorer | Bax channel blocker |
For Research Use Only