Läs mer
BAX Mouse anti-Human, CoraLite 488, Clone: 4G5E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60267-100UL
Beskrivning
The BAX antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human BAX. This antibody recognizes human, mouse, rat antigen. The BAX antibody has been validated for the following applications: FC analysis. This antibody is coralite™488.
Specifikationer
| BAX | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 581 | |
| IgG2b |
| Western Blot, Flow Cytometry | |
| 4G5E8 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.