Läs mer
BAY 11-7085, TRC
Produktkod. 30240526
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240526 25 mg
30240964 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240526 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB11879525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 196309-76-9
Molekylformel C13H15NO2S
Molekylvikt (g/mol) 249.33
Synonym BAY 11-7085, (E)-3-[[4-(1,1-Dimethylethyl)phenyl]sulfonyl]-2-propenenitrile, BAY 117083
IUPAC-namn (E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile
LEDER CC(C)(C)c1ccc(cc1)S(=O)(=O)\C=C\C#N
Specifikationer
Kemiskt namn eller material (2E)-3-[(4-(1,1-Dimethylethyl)phenyl)sulfonyl]-2-propenenitrile
Kvantitet 25 mg
InChI formel InChI=1S/C13H15NO2S/c1-13(2,3)11-5-7-12(8-6-11)17(15,16)10-4-9-14/h4-8,10H,1-3H3/b10-4+