Läs mer
BAY-1895344, TRC
Produktkod. 30240680
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240680 1 mg
30240675 10 mg
30240531 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240680 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1189251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1876467-74-1
Molekylformel C20H21N7O
Synonym 2-[(3R)-3-Methyl-4-morpholinyl]-4-(1-methyl-1H-pyrazol-5-yl)-8-(1H-pyrazol-3-yl)-1,7-naphthyridine∣BAY-1895344
Specifikationer
Kemiskt namn eller material (R)-3-Methyl-4-(4-(1-methyl-1H-pyrazol-5-yl)-8-(1H-pyrazol-5-yl)-1,7-naphthyridin-2-yl)morpholine
Kvantitet 1 mg
Frakt skick Room Temperature
MDL-nummer MFCD31561442
Molekylvikt (g/mol) 375.43, 375.44
InChI-nyckel YBXRSCXGRPSTMW-CYBMUJFWSA-N
LEDER C[C@@H]1COCCN1C1=NC2=C(N=CC=C2C(=C1)C1=CC=NN1C)C1=CC=NN1
IUPAC-namn 4-(1-methyl-1H-pyrazol-5-yl)-2-[(3R)-3-methylmorpholin-4-yl]-8-(1H-pyrazol-5-yl)-1,7-naphthyridine