missing translation for 'onlineSavingsMsg'
Learn More

BAY-293, MedChemExpress

Product Code. 16485616
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
16485616 100 mg
16441087 50 mg
16434828 10 mg
16427577 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485616 Supplier Medchem Express Supplier No. HY114398

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY-293, a valuable chemical probe, blocks RAS activation via disruption of the KRAS-SOS1 interaction with an IC50 of 21 nM. BAY-293 is a potent inhibitor of Son of Sevenless 1 (SOS1). SOS1 is the guanine nucleotide exchange factor (GEF) and activator of RAS.

BAY-293, a valuable chemical probe, blocks RAS activation via disruption of the KRAS-SOS1 interaction with an IC50 of 21 nM. BAY-293 is a potent inhibitor of Son of Sevenless 1 (SOS1). SOS1 is the guanine nucleotide exchange factor (GEF) and activator of RAS.

Specifications

Kemiskt namn eller material BAY-293
CAS 2244904-70-7
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C25H28N4O2S
Kvantitet 100 mg
Löslighetsinformation DMSO : ≥ 125 mg/mL (278.66 mM)
LEDER COC1=C(OC)C=C(N=C(C)N=C2N[C@H](C)C3=CC(C4=C(CNC)C=CC=C4)=CS3)C2=C1
Molekylvikt (g/mol) 448.58
Formel vikt 448.58
Procent renhet 98.06%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.