Läs mer
Tocris Bioscience™ BAY 58-2667 hydrochloride, Tocris Bioscience™
Potent soluble guanylyl cyclase (sGC) activator
Brand: Tocris Bioscience™ 6052/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
BAY 58-2667 hydrochloride is a potent soluble guanylyl cyclase (sGC) activator (EC50 = 6.4 nM). Binds heme-free sGC. Exhibits antihypertensive effects in vivo.Specifikationer
| BAY 58-2667 hydrochloride | |
| 4-[[(4-Carboxybutyl)[2-[2-[[4-(2-phenylethyl)phenyl]methoxy]phenyl]ethyl]amino]methyl]benzoic acid hydrochloride | |
| Soluble to 100mM in DMSO | |
| White |
| 646995-35-9 | |
| 10 mg | |
| Solid | |
| Bioactive Small Molecule |