missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ BAY 598
GREENER_CHOICE

Produktkod. 18758286
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18758286 25 mg
18757456 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18758286 Leverantör Tocris Bioscience™ Leverantörsnummer 5991/25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Potent and selective SMYD2 inhibitor

BAY 598 is a potent and selective competitive inhibitor of SMYD2 lysine methyltransferase (IC50 values are 27 and 58 nM for biochemical and cellular activity assays, respectively), which displays >100-fold selectivity for SMYD2 over a panel of 32 other methyltransferases including SMYD3, SUV420H1, and SUV420H2. BAY 598 enhances apoptotic responses to doxorubicin (Cat. No. 2252) in cancer cell lines, and decreases p53K370me levels in HEK293 cells. It also reduces methylation in tumor cells in a mouse xenograft model.

To request the negative control for BAY 598, please fill out the BAY 369 request form on the SGC website.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material N-[(4 S)-1-[(Cyanoamino)[[3-(difluoromethoxy)phenyl]imino]methyl]-3-(3,4-dichlorophenyl)-4,5-dihydro-1 H-pyrazol-4-yl]-N-ethyl-2-hydroxyacetamide
CAS 1906919-67-2
Kvantitet 25 mg
Mål Additional Lysine Methyltransferase Compounds
Molekylformel C22H20Cl2F2N6O3
Formel vikt 525.34
Renhet 98%
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.