Läs mer
BAY 677
Produktkod. 18791839
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18791839 25 mg
18758078 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18791839 Leverantör Tocris Bioscience Leverantörsnummer 6389/25
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Inactive control for BAY 678 (Cat.No.5706)
BAY 677 is an inactive control for BAY 678 (Cat.No.5706).
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-[(4S)-5-Acetyl-1,2,3,4-tetrahydro-6-methyl-2-oxo-1-[3-(trifluoromethyl)phenyl]-4-pyrimidinyl]-2-pyridinecarbonitrile
CAS 2117404-84-7
Analysprocentintervall >98%
Kvantitet 25 mg
Molekylformel C20H15F3N4O2
Formel vikt Observed MW: 400.35