Läs mer
BAY 73-6691, MedChemExpress
Produktkod. 16493928
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16493928 10 mg
16476647 5 mg
16470287 50 mg
16454956 100 mg
16442916 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16493928 Leverantör Medchem Express Leverantörsnummer HY104028
MedChemExpress BAY 73-6691 ((R)-BAY 73-6691) is a potent, brain penetrant, and selective PDE9A inhibitor.
BAY 73-6691 ((R)-BAY 73-6691) is a potent, brain penetrant, and selective PDE9A inhibitor.
Specifikationer
Kemiskt namn eller material BAY 73-6691
Fysisk form Solid
CAS 794568-92-6
Färg Light Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C15H12ClF3N4O
För användning med (applikation) Neuroscience-Neurodegeneration
Synonym (R)-BAY 73-6691
Löslighetsinformation DMSO : 160 mg/mL (448.52 mM; Need ultrasonic and warming)
LEDER O=C1C2=C(N(C3=CC=CC=C3Cl)N=C2)NC(C[C@@H](C)C(F)(F)F)=N1
Molekylvikt (g/mol) 356.73
Formel vikt 356.73
Procent renhet 99.81%