Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepared Media
Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens
606.00 SEK - 2350.00 SEK
Specifications
Color | Beige |
---|---|
Content And Storage | 15°C to 25°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Trypticase Soy Broth |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
17332321
|
BD
221093 |
Size K tubes with cap | 100 x 8 mL |
1695.00 SEK
Pack of 100 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
16512382
|
BD
221815 |
Size K tubes with cap | 100 x 2 mL |
1355.00 SEK
Pack of 100 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
17352341
|
BD
221716 |
Size K tubes with cap | 100 x 5 mL |
1660.00 SEK
Pack of 100 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
15142265
|
BD
297354 |
Size K tubes with cap | 100 x 10 mL |
2350.00 SEK
10mL |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
17322321
|
BD
221092 |
Size K tubes with cap | 10 x 8 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Beige | |
Liquid | |
Trypticase Soy Broth | |
For qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms |
15°C to 25°C | |
United States | |
Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L |
For in vitro diagnostic use.