missing translation for 'onlineSavingsMsg'
Learn More

BBS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21118-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840725

  • 3313.85 SEK / 150µL
Estimated Shipment: 03-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mutations in this gene have been observed in patients with the major form (type 1) of Bardet-Biedl syndrome. The encoded protein may play a role in eye, limb, cardiac and reproductive system development.
Specifications

Specifications

BBS1
Polyclonal
Unconjugated
BBS1
Bardet Biedl syndrome 1, BBS1, BBS2 like protein 2, BBS2L2
Rabbit
Antigen Affinity Chromatography
RUO
52028, 582
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3V3N7, Q8NFJ9
Bbs1
BBS1 Fusion Protein Ag15460
150 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers