missing translation for 'onlineSavingsMsg'
Learn More

BBS5 Mouse anti-Human, Mouse, Clone: 4G8E12, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66136-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16897735

  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The BBSome complex is thought to function as a coat complex required for sorting of specific membrane proteins to the primary cilia. The BBSome complex is required for ciliogenesis but is dispensable for centriolar satellite function. This ciliogenic function is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome. Rab8(GTP) enters the primary cilium and promotes extension of the ciliary membrane. Firstly the BBSome associates with the ciliary membrane and binds to RAB3IP/Rabin8, the guanosyl exchange factor (GEF) for Rab8 and then the Rab8-GTP localizes to the cilium and promotes docking and fusion of carrier vesicles to the base of the ciliary membrane. The BBSome complex, together with the LTZL1, controls SMO ciliary trafficking and contributes to the sonic hedgehog (SHH) pathway regulation. Required for BBSome complex ciliary localization but not for the proper complex assembly.
Specifications

Specifications

BBS5
Monoclonal
0.72 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N3I7, Q9CZQ9
Bbs5
BBS5 Fusion Protein Ag6844
20 μL
Primary
Mouse, Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
4G8E12
Unconjugated
BBS5
Bardet Biedl syndrome 5, BBS5
Mouse
Protein G
RUO
129880, 72569
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers