missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS8 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | BBS8 |
---|---|
Concentration | 353 μg/mL |
Applications | Western Blot, Immunoprecipitation, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850254
|
Proteintech
12505-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840254
|
Proteintech
12505-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The BBS8 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BBS8. This antibody recognizes human, mouse, rat antigen. The BBS8 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
BBS8 | |
Western Blot, Immunoprecipitation, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
353 μg/mL | |
Polyclonal | |
Rabbit | |
123016 | |
Primary |