Läs mer
Tocris Bioscience™ BC-LI-0186
Beskrivning
BC-LI-0186 is a potent and high affinity leucyl-tRNA synthase (TRS)/Ras-related GTP-binding protein D (RagD) interaction inhibitor (IC50 = 46.1 nM; Kd = 42.1 nM for binding to LRS). Inhibits mTOR complex 1 activity by blocking binding to LRS without affecting catalytic activity. Inhibits leucine-induced translocation of LRS to the lysosome.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-[2,5-Dihydro-2,3-dimethyl-4-(1-methylethyl)-5-oxo-1 H-pyrazol-1-yl]-N-(2-phenoxyethyl)benzenesulfonamide |
| CAS | 695207-56-8 |
| Kvantitet | 25 mg |
| Mål | Ras GTPase Inhibitors |
| Molekylformel | C22H27N3O4S |
| Formel vikt | 3939 |
| Renhet | 98% |