missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10402-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
BCAS3 is physically associated with histone H3 and histone acetyltransferase complex protein P/CAF (p300/CBP-associated factor) and possesses histone acetyltransferase activity. In breast cancer, several chromosomal sites frequently undergo amplification, implicating the location of genes important for tumor development and progression.Specifications
BCAS3 | |
Polyclonal | |
Unconjugated | |
BCAS3 | |
BCAS3, GAOB1, MAAB, Protein Maab1, Rudhira | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192197, 363662, 54828 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CCN5, Q9H6U6 | |
Bcas3 | |
BCAS3 Fusion Protein Ag0646 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |