Läs mer
MedChemExpress BCECF
Specifications
Specifications
| Innehåll och lagring | -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Produkttyp | Dye Reagents |
| Kvantitet | 1 mg |
| Frakt skick | Room temperature |
| Löslighet | DMSO : 62.5 mg/mL (120.09 mM; Need ultrasonic) |
| Molekylvikt (g/mol) | 520.44 g/mol |
| CAS | 85138-49-4 |
| Synonym | 2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein |