missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCHE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23854-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Mutant alleles at the BCHE locus are responsible for suxamethonium sensitivity. Homozygous persons sustain prolonged apnea after administration of the muscle relaxant suxamethonium in connection with surgical anesthesia. The activity of pseudocholinesterase in the serum is low and its substrate behavior is atypical. In the absence of the relaxant, the homozygote is at no known disadvantage.Specifications
BCHE | |
Polyclonal | |
Unconjugated | |
BCHE | |
Acylcholine acylhydrolase, BCHE, Butyrylcholine esterase, butyrylcholinesterase, CHE1, Choline esterase II, Cholinesterase, E1, Pseudocholinesterase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
590 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06276 | |
BCHE | |
BCHE Fusion Protein Ag20888 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |