missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ BCKDHA (Human) Recombinant Protein

Human BCKDHA full-length ORF ( AAH07878.1, 1 a.a. - 445 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000593-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16172151

  • 4515.00 SEK / 10µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MAVAIAAARVWRLNRGLSQAALLLLRQPGARGLARSHPPRQQQQFSSLDDKPQFPGASAEFIDKLEFIQPNVISGIPIYRVMDRQGQIINPSEDPHLPKEKVLKLYKSMTLLNTMDRILYESQRQGRISFYMTNYGEEGTHVGSAAALDNTDLVFGQYREAGVLMYRDYPLELFMAQCYGNISDLGKGRQMPVHYGCKERHFVTISSPLATQIPQAVGAAYAAKRANANRVVICYFGEGAASEGDAHAGFNFAATLECPIIFFCRNNGYAISTPTSEQYRGDGIAARGPGYGIMSIRVDGNDVFAVYNATKEARRRAVAENQPFLIEAMTYRIGHHSTSDDSSAYRSVDEVNYWDKQDHPISRLRHYLLSQGWWDEEQEKAWRKQSRRKVMEAFEQAERKPKPNPNLLFSDVYQEMPAQLRKQQESLARHLQTYGEHYPLDHFDK
Specifications

Specifications

AAH07878.1
branched chain keto acid dehydrogenase E1, alpha polypeptide
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCKDHA
GST
593
Wheat germ expression system
10 μg
BCKDE1A, FLJ45695, MSU, MSUD1, OVD1A
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers