missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Bcl-2 Monoclonal Antibody (Bcl-2-100)
GREENER_CHOICE

Produktkod. 10671034
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
Varje
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
10671034 100 μg Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy

For Research Use Only. All usage must comply with product instructions.

Produktkod. 10671034 Leverantör Invitrogen™ Leverantörsnummer 138800

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

For Research Use Only. All usage must comply with product instructions.

Mouse Monoclonal Antibody

Bcl-2 Monoclonal Antibody for Flow, IHC, IHC (P), IP, Western Blot

BCL-2 is a key regulator of apoptosis that functions to either inhibit or promote cell death. The BCL-2 family members are also characterized by dimerizing to further modulate apoptosis. Bag1, for example, has been found to form a heterodimer with BCL-2 resulting in the enhancement of the anti-apoptotic effect of BCL-2. Bax and Bak have been shown to play a critical role in cytochrome c release from mitochondria and thus initiate apoptosis. Bax exerts a pro-apoptotic rather than an anti-apoptotic effect on cells. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. In most follicular lymphomas, neoplastic germinal centers express high levels of BCL-2 alpha protein, whereas the normal or hyperplastic germinal centers are negative. Two transcript variants of BCL-2, produced by alternate splicing, differ in their C-terminal ends. The overexpression of BCL-2 has been linked to human cancers such as B-cell lymphoma and prostate cancer.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Bcl-2
Användningsområden Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Klassificering Monoclonal
Klona Bcl-2-100
Koncentration 0.5 mg/mL
Konjugera Unconjugated
Formulering PBS with 0.1% sodium azide; pH 7.4
Gen Bcl2
Genaccessionsnr. P10415, P10417
Gene Alias apoptosis regulator Bcl-2; AW986256; B cell leukemia/lymphoma 2; B cell lymphoma 2 associated oncogene; B-cell CLL/lymphoma 2; B-cell leukemia/lymphoma 2; Bcl 2; BCL2; bcl-2; BCL2 apoptosis regulator; Bcl-2 protein; BCL2, apoptosis regulator; Bcl2-like protein; C430015F12Rik; D630044D05Rik; D830018M01Rik; OTTHUMP00000163680; PPP1R50; protein phosphatase 1, regulatory subunit 50
Gensymboler Bcl2
Värd art Mouse
Immunogen Synthetic peptide corresponding to residues 41-54 of the human Bcl-2 protein. This sequence is not shared by the mouse Bcl-2 protein.
Reningsmetod Affinity chromatography
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 12043, 596
Målarter Human, Mouse
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG1 κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.