missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Bcl-2 Recombinant Rabbit Monoclonal Antibody (2C6)
GREENER_CHOICE

Produktkod. 16555751
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16555751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16555751 Leverantör Invitrogen™ Leverantörsnummer MA533119

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

Bcl-2 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

BCL-2 is a key regulator of apoptosis that functions to either inhibit or promote cell death. The BCL-2 family members are also characterized by dimerizing to further modulate apoptosis. Bag1, for example, has been found to form a heterodimer with BCL-2 resulting in the enhancement of the anti-apoptotic effect of BCL-2. Bax and Bak have been shown to play a critical role in cytochrome c release from mitochondria and thus initiate apoptosis. Bax exerts a pro-apoptotic rather than an anti-apoptotic effect on cells. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. In most follicular lymphomas, neoplastic germinal centers express high levels of BCL-2 alpha protein, whereas the normal or hyperplastic germinal centers are negative. Two transcript variants of BCL-2, produced by alternate splicing, differ in their C-terminal ends. The overexpression of BCL-2 has been linked to human cancers such as B-cell lymphoma and prostate cancer.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Bcl-2
Användningsområden ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Klassificering Recombinant Monoclonal
Klona 2C6
Koncentration 0.4 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen Bcl2
Genaccessionsnr. P10415, P10417, P49950
Gene Alias apoptosis regulator Bcl-2; AW986256; B cell leukemia/lymphoma 2; B cell lymphoma 2 associated oncogene; B-cell CLL/lymphoma 2; B-cell leukemia/lymphoma 2; Bcl 2; BCL2; bcl-2; BCL2 apoptosis regulator; Bcl-2 protein; BCL2, apoptosis regulator; Bcl2-like protein; C430015F12Rik; D630044D05Rik; D830018M01Rik; OTTHUMP00000163680; PPP1R50; protein phosphatase 1, regulatory subunit 50
Gensymboler Bcl2
Värd art Rabbit
Immunogen Synthesized Bcl-2 Antibody (48-81aa).
Reningsmetod Antigen Affinity Chromatography, Protein G
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 12043, 24224, 596
Målarter Human, Mouse, Rat
Innehåll och lagring -20°C or -80°C if preferred
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.