missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22947-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
BCL9 is involved in signal transduction through the Wnt pathway. It promotes beta-catenin′s transcriptional activity. BCL9 is associated with B-cell acute lymphoblastic leukemia. The overexpression of BCL9 may be of pathogenic significance in B-cell maligciesSpecifications
BCL9 | |
Polyclonal | |
Unconjugated | |
BCL9 | |
B cell CLL/lymphoma 9, B cell CLL/lymphoma 9 protein, B cell lymphoma 9 protein, BCL9, LGS, Protein legless homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
607 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00512 | |
BCL9 | |
BCL9 Fusion Protein Ag19158 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |