missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | BCL9 |
---|---|
Concentration | 0.44 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16822145
|
Proteintech
22947-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16812145
|
Proteintech
22947-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
BCL9 is involved in signal transduction through the Wnt pathway. It promotes beta-catenin′s transcriptional activity. BCL9 is associated with B-cell acute lymphoblastic leukemia. The overexpression of BCL9 may be of pathogenic significance in B-cell maligciesSpecifications
BCL9 | |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
O00512 | |
607 | |
BCL9 Fusion Protein Ag19158 | |
Primary | |
-20°C | |
BCL9 |
0.44 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B cell CLL/lymphoma 9, B cell CLL/lymphoma 9 protein, B cell lymphoma 9 protein, BCL9, LGS, Protein legless homolog | |
BCL9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |