missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
BCR Mouse anti-Human, Clone:1E11G12, Abnova™
Beskrivning
A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifikationer
Specifikationer
| Antigen | BCR |
| Användningsområden | ELISA, Flow Cytometry, Western Blot |
| Klassificering | Monoclonal |
| Klona | 1E11G12 |
| Konjugera | Unconjugated |
| Beskrivning | breakpoint cluster region |
| Utspädning | ELISA (1:10000) Flow Cytometry (1:200-1:400) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.05% sodium azide) |
| Gen | BCR |
| Gene Alias | ALL, BCR-ABL1, BCR1, CML, D22S11, D22S662, FLJ16453, PHL |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?