missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Bcr Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16643755
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16643755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16643755 Leverantör Invitrogen™ Leverantörsnummer PA599505

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Bcr.

A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Bcr
Användningsområden Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen Bcr
Genaccessionsnr. P11274, Q6PAJ1
Gene Alias 5133400C09Rik; ABL; AI561783; AI853148; ALL; BCR; BCR activator of RhoGEF and GTPase; BCR protein; BCR, RhoGEF and GTPase activating protein; bcr/abl; BCR/FGFR1 chimera protein; BCR1; breakpoint cluster region; breakpoint cluster region homolog; breakpoint cluster region protein; c-ABL; c-abl oncogene 1; CML; D22S11; D22S662; EC 2.7.11.1; FGFR1/BCR chimera protein; JTK7; Kiaa3017; mKIAA3017; p150; PHL; renal carcinoma antigen NY-REN-26; RP11-83J21.1; v-abl
Gensymboler Bcr
Värd art Rabbit
Immunogen A synthesized peptide derived from human BCR(Accession P11274), corresponding to amino acid residues V151-S201.
Reningsmetod Affinity Chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 110279, 613
Målarter Human, Mouse
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.