Läs mer
BD Apoptosis I Sampler Kit
For study of programmed cell death
Brand: BD 612741
Varning:
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Innehåller: Apaf-1, Bad, Bax, Bcl-2, Bcl-x, BRUCE, CAS, hILP/XIAP, Nip1, p53, Storage Buffer: aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide
Beskrivning
Apoptosis is a selective process of genetically programmed cell death which occurs during normal cellular differentiation and development of multicellular organisms. This process is also involved in tissue homeostasis, pathological conditions and aging.
- Concentration: 250μg/mL
- Format: Purified
Western Blotting
Specifikationer
| 1 Kit |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Research use only