Learn More
BD BBL™ Mycoflask™ Malonate Broth (Ewing Modified) Prepared Media
For the differentiation of coliforms and other enteric organisms
Brand: BD 221322
Additional Details : CAS Number : 7647-14-5 Weight : 0.50000kg
Specifications
CE, AOAC, BAM, COMPF, SMD | |
Liquid | |
Yeast extract 1g/L, Ammonium Sulfate 2g/L, Dipotassium Phosphate 0.6g/L, Monopotassium Phosphate 0.4g/L, Sodium Chloride 2g/L, Sodium Malonate 3g/L, Dextrose 0.25g/L, Bromthymol blue 0.025g/L | |
Tube with Tight Cap |
Green | |
United States | |
For differentiation of Coliforms and other enteric organisms | |
5 mL |
For in vitro Diagnostic Use.