missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Middlebrook and Cohn 7H10 Prepared Agar Slants
Prepared media. BBL™ Mycoflask™ Middlebrook and Cohn 7H10 Prepared Agar Slants are used in qualitative procedures for the isolation and cultivation of mycobacteria.
Brand: BD 220959
Additional Details : Weight : 0.01000kg
Description
- Contains a variety of inorganic salts that provide substances essential for the growth of mycobacteria
Specifications
ISO/DIS, 11014, BS12, CMPH2, EP, MCM9 | |
2°C to 8°C | |
United States | |
Magnesium Sulfate 0.05g/L, Ferric Ammonium Citrate 0.04g/L, Sodium Citrate 0.4g/L, Ammonium Sulfate 0.5g/L, Monosodium Glutamate 0.5g/L, Disodium Phosphate 1.5g/L, Monopotassium Phosphate 1.5g/L, Agar 13.5g/L, Sodium Chloride 0.85g/L, Dextrose 2g/L, Bovine Albumin (Fraction V) 5g/L, Catalase 3mg/L, Pyridoxine 1mg/L, Zinc Sulfate 1mg/L, Copper Sulfate 1mg/L, Biotin 0.5mg/L, Calcium Chloride 0.5mg/L, Malachite green 0.25mg/L, Oleic Acid 0.06mLg/L, Glycerol 5mLg/L | |
Tube |
Gray | |
Highly Viscous | |
Middlebrook and Cohn 7H10 Agar | |
Qualitative procedures for the isolation and cultivation of mycobacteria | |
100 Pack |
For in vitro Diagnostic Use.