Learn More
Beschreibung
Beinhaltet
Reagents for capture and detection of IL-8, RANTES, MIG, MCP-1, and IP-10, wash buffer, assay diluent, cytometer setup reagents
- Uses the sensitivity of amplified fluorescence detection by flow cytometry to measure soluble analytes in a particle-based immunoassay
- Each bead in the kit provides a capture surface for a specific protein and is analogous to an individually coated well in an ELISA plate
- The Capture Bead mixture is in suspension to allow for the detection of multiple analytes in a small volume sample
- The broad dynamic range of fluorescent detection via flow cytometry and the efficient capturing of analytes via suspended particles enable assay to use fewer sample dilutions and to determine the concentration of an unknown analyte in substantially less time (compared to conventional ELISA)
- Can be used to quantitatively measure Interleukin-8 (CXCL8/IL-8), RANTES (CCL5/RANTES), Monokine-induced by Interferon-γ (CXCL9/MIG), Monocyte Chemoattractant Protein-1 (CCL2/MCP-1), and Interferon-γ-induced Protein-10 (CXCL10/IP-10) levels in a single sample
- Provides sufficient reagents for the quantitative analysis of 80 samples
For research use only. Not for use in diagnostic or therapeutic procedures.
Spezifikation
Spezifikation
| Detects | IL-8, RANTES, MIG, MCP-1 and IP-10 |
| For Use With (Application) | For analysis of specific chemokines in Tissue Culture supernatants, EDTA-treated Plasma (EDTA-plasma) and Serum samples |
| Quantity | 80 Tests |
| Content And Storage | 2°C to 8°C |
| Reactivity | Human |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?