missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Luria Agar Base, Miller

For propagation and maintenance of E. coli for use in molecular biology

Brand:  BD 241320

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11748862

  • 1981.90 SEK / 500g

Estimated Shipment
12-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Contains 0.5g/L NaCl

Applications:

For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria

Specifications

Specifications

Pancreatic digest of Casein 10g/L, Yeast extract 5g/L, Sodium Chloride 0.5g/L, Agar 15g/L
Luria Agar Base
1825 Days
Dehydrated
CE
Refrigerate
Maintaining and propagating Escherichia coli in molecular microbiology procedures with or without added Glucose
500g
Luria Agar Base
Bottle
Videos
SDS
Documents